Merck
  • Benzophenone derivatives of pyrimidines as effective non-nucleoside inhibitors of wild-type and drug-resistant HIV-1 reverse transcriptase.
Benzophenone derivatives of pyrimidines as effective non-nucleoside inhibitors of wild-type and drug-resistant HIV-1 reverse transcriptase.
Doklady. Biochemistry and biophysics (2013-01-05)
M M Prokofjeva, V T Valuev-Elliston, A V Ivanov, S N Kochetkov, M S Novikov, V S Prassolov
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzophenon, ReagentPlus®, 99%
Supelco
Benzophenon, analytical standard
Supelco
Mettler-Toledo-Kalibriersubstanz ME 18870, Benzophenon, traceable to primary standards (LGC)
Supelco
Benzophenon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzophenon, purified by sublimation, ≥99%
Supelco
Schmelzpunktstandard 47-49°C, analytical standard
Benzophenon, European Pharmacopoeia (EP) Reference Standard