Merck
  • Meta-analysis of carvedilol versus beta 1 selective beta-blockers (atenolol, bisoprolol, metoprolol, and nebivolol).
Meta-analysis of carvedilol versus beta 1 selective beta-blockers (atenolol, bisoprolol, metoprolol, and nebivolol).
The American journal of cardiology (2013-01-08)
James J DiNicolantonio, Carl J Lavie, Hassan Fares, Arthur R Menezes, James H O'Keefe
ZUSAMMENFASSUNG
Because carvedilol is a unique vasodilating β blocker (BB) exerting antioxidant activity and pleiotropic effects, it was theorized that it may confer more potent beneficial effects on cardiovascular mortality and morbidity in acute myocardial infarction (AMI) and heart failure (HF) settings. A systematic review and meta-analysis was performed of randomized, controlled, direct-comparison trials that included adults receiving atenolol, bisoprolol, metoprolol, nebivolol, or carvedilol to evaluate the effects of carvedilol compared to other BBs on mortality, cardiovascular events, and hospital readmissions in the setting of AMI or systolic HF. Compared to β(1)-selective BBs used in HF (8 trials, n = 4,563), carvedilol significantly reduced all-cause mortality (risk ratio 0.85, 95% confidence interval 0.78 to 0.93, p = 0.0006). In 3 trials of patients with AMI (n = 644), carvedilol significantly reduced all-cause mortality by 45% (fixed-effects model: risk ratio 0.55, 95% confidence interval 0.32 to 0.94, p = 0.03, random-effects model: risk ratio 0.56, 95% confidence interval 0.26 to 1.12, p = 0.10), with no reduction in non-fatal MI (risk ratio 0.61, 95% confidence interval 0.31 to 1.22, p = 0.16). In conclusion, carvedilol, as compared against atenolol, bisoprolol, metoprolol and nebivolol in randomized direct comparison trials, significantly reduced all-cause mortality in systolic HF patients. Additionally, carvedilol significantly reduced all-cause mortality compared with β(1)-selective BBs in AMI patients using the fixed-effects model but not using the random-effects model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
(±)-Metoprolol, ≥98% (titration), powder
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Supelco
Metoprolol-Tartrat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Metoprolol tartrate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Bisoprolol -hemifumarat (Salz), ≥98% (HPLC), solid
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Bisoprololfumarat, European Pharmacopoeia (EP) Reference Standard
Bisoprolol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
USP
Carvedilol, United States Pharmacopeia (USP) Reference Standard
Bisoprolol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Carvedilol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Metoprolol -tartrat, European Pharmacopoeia (EP) Reference Standard
Atenolol, European Pharmacopoeia (EP) Reference Standard
Carvedilol, European Pharmacopoeia (EP) Reference Standard