Merck
  • Melatonin enhances DNA repair capacity possibly by affecting genes involved in DNA damage responsive pathways.
Melatonin enhances DNA repair capacity possibly by affecting genes involved in DNA damage responsive pathways.
BMC cell biology (2013-01-09)
Ran Liu, Alan Fu, Aaron E Hoffman, Tongzhang Zheng, Yong Zhu
ZUSAMMENFASSUNG
Melatonin, a hormone-like substance involved in the regulation of the circadian rhythm, has been demonstrated to protect cells against oxidative DNA damage and to inhibit tumorigenesis. In the current study, we investigated the effect of melatonin on DNA strand breaks using the alkaline DNA comet assay in breast cancer (MCF-7) and colon cancer (HCT-15) cell lines. Our results demonstrated that cells pretreated with melatonin had significantly shorter Olive tail moments compared to non-melatonin treated cells upon mutagen (methyl methanesulfonate, MMS) exposure, indicating an increased DNA repair capacity after melatonin treatment. We further examined the genome-wide gene expression in melatonin pretreated MCF-7 cells upon carcinogen exposure and detected altered expression of many genes involved in multiple DNA damage responsive pathways. Genes exhibiting altered expression were further analyzed for functional interrelatedness using network- and pathway-based bioinformatics analysis. The top functional network was defined as having relevance for "DNA Replication, Recombination, and Repair, Gene Expression, [and] Cancer". These findings suggest that melatonin may enhance DNA repair capacity by affecting several key genes involved in DNA damage responsive pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylmethansulfonat, 99%
Supelco
Methylmethansulfonat, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland