Merck
  • Adverse reactions following mass drug administration with diethylcarbamazine in lymphatic filariasis endemic areas in the Northeast of Brazil.
Adverse reactions following mass drug administration with diethylcarbamazine in lymphatic filariasis endemic areas in the Northeast of Brazil.
Revista da Sociedade Brasileira de Medicina Tropical (2013-01-09)
Ana Wládia Lima, Zulma Medeiros, Zailde Carvalho Dos Santos, Gertrudes Monteiro da Costa, Cynthia Braga
ZUSAMMENFASSUNG
The Global Programme to Eliminate Lymphatic Filariasis was launched with the goal of eliminating this disease via the annual mass drug administration (MDA) of a single dose of antifilarial drugs. Adverse drug reactions following MDA are a major factor of poor treatment adherence in several countries. This study assessed the occurrence of adverse drug reactions (ADRs) following the first round of mass treatment in two communities treated with different dosages of diethylcarbamazine (DEC) in the City of Recife, Brazil. Population-based cross-sectional surveys were conducted in a random sample of the population living in both communities (Areas I and II). The dose of DEC recommended by the WHO (6mg/kg) was calculated based on the individual's weight-for-age. In Area II, weight differences between the genders were also considered when determining dosage. Data were obtained through interviews conducted in the first 12 to 48h and on the 5th day after MDA during household visits. A total of 487 and 365 individuals were interviewed in Areas I and II, respectively. The prevalence of ADRs in Area I (23.6; 95%CI: 19.1-29.5) was higher than in Area II (16.2; 95%CI:11.9-21.5)(p=0.0078). The prevalence of ADRs among females was higher than in males in Area I (p=0.0021). In Area II, no significant difference between the genders was observed (p=0.1840). Age was not associated with ADRs in either area. Adjusting MDA dosage schedules according to weight-for-age and sex may be may contribute to reduce the occurrence of adverse drug reactions in the population.