Merck
  • The relationships between pesticide metabolites and neurobehavioral test performance in the third National Health and Nutrition Examination Survey.
The relationships between pesticide metabolites and neurobehavioral test performance in the third National Health and Nutrition Examination Survey.
Archives of environmental & occupational health (2013-01-10)
Edward F Krieg
ZUSAMMENFASSUNG
Regression analysis was used to estimate and test for relationships between urinary pesticide metabolites and neurobehavioral test performance in adults, 20 to 59 years old, participating in the third National Health and Nutrition Examination Survey. The 12 pesticide metabolites included 2 naphthols, 8 phenols, a phenoxyacetic acid, and a pyridinol. The 3 neurobehavioral tests included in the survey were simple reaction time, symbol-digit substitution, and serial digit learning. As the 2,4-dichlorophenol, 2,5-dichlorophenol, and the pentachlorophenol concentrations increased, performance on the serial digit learning test improved. As the 2,5-dichlorophenol concentration increased, performance on the symbol-digit substitution test improved. At low concentrations, the parent compounds of these metabolites may act at acetylcholine and γ-aminobutyric acid synapses in the central nervous system to improve neurobehavioral test performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,4-Dichlorphenoxyessigsäure, 97%
Sigma-Aldrich
2,4-Dichlorphenoxyessigsäure, ≥95%, crystalline
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
3,5,6-Trichlor-2-pyridinol, PESTANAL®, analytical standard