Merck
  • Enhanced SH3/linker interaction overcomes Abl kinase activation by gatekeeper and myristic acid binding pocket mutations and increases sensitivity to small molecule inhibitors.
Enhanced SH3/linker interaction overcomes Abl kinase activation by gatekeeper and myristic acid binding pocket mutations and increases sensitivity to small molecule inhibitors.
The Journal of biological chemistry (2013-01-11)
Shoghag Panjarian, Roxana E Iacob, Shugui Chen, Thomas E Wales, John R Engen, Thomas E Smithgall
ZUSAMMENFASSUNG
Multidomain kinases such as c-Src and c-Abl are regulated by complex allosteric interactions involving their noncatalytic SH3 and SH2 domains. Here we show that enhancing natural allosteric control of kinase activity by SH3/linker engagement has long-range suppressive effects on the kinase activity of the c-Abl core. Surprisingly, enhanced SH3/linker interaction also dramatically sensitized the Bcr-Abl tyrosine kinase associated with chronic myelogenous leukemia to small molecule inhibitors that target either the active site or the myristic acid binding pocket in the kinase domain C-lobe. Dynamics analyses using hydrogen exchange mass spectrometry revealed a remarkable allosteric network linking the SH3 domain, the myristic acid binding pocket, and the active site of the c-Abl core, providing a structural basis for the biological observations. These results suggest a rational strategy for enhanced drug targeting of Bcr-Abl and other multidomain kinase systems that use multiple small molecules to exploit natural mechanisms of kinase control.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
X-tremeGENE 9 DNA-Transfektionsreagenz, Polymer reagent for transfecting common cell lines
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Myristinsäure, natural, ≥98.5%, FG
Sigma-Aldrich
Myristinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Myristinsäure, ≥98.0% (GC)
Supelco
Myristinsäure, analytical standard
Supelco
Myristinsäure, Pharmaceutical Secondary Standard; Certified Reference Material