Merck
  • Production and partial characterization of cellulases and Xylanases from Trichoderma atroviride 676 using lignocellulosic residual biomass.
Production and partial characterization of cellulases and Xylanases from Trichoderma atroviride 676 using lignocellulosic residual biomass.
Applied biochemistry and biotechnology (2013-01-12)
André Luiz Grigorevski-Lima, Mariana Menezes Quadros de Oliveira, Rodrigo Pires do Nascimento, Elba Pinto da Silva Bon, Rosalie Reed Rodrigues Coelho
ZUSAMMENFASSUNG
Trichoderma atroviride 676 was studied to evaluate its efficiency in the production of some lignocellulolytic enzymes, using lignocellulosic residual biomass. Best results were obtained when 3.0 % (w/v) untreated sugarcane bagasse was used (61.3 U mL(-1) for xylanase, 1.9 U mL(-1) for endoglucanase, 0.25 U mL(-1) for FPase, and 0.17 U mL(-1) for β-glucosidase) after 3-4 days fermentation. The maximal enzymatic activity for endoglucanase, FPase, and xylanase were observed at 50-60 °C and pH 4.0-5.0, whereas thermal stability at 50 °C (CMCase and FPase) or 40 °C (xylanase) was obtained after 8 h. Zymograms have shown two bands of 104 and 200 kDa for endoglucanases and three bands for xylanase (23, 36, and 55.7 kDa). The results obtained with T. atroviride strain 676 were comparable to those obtained with the cellulolytic strain Trichoderma reesei RUT-C30, indicating, in the studied conditions, its great potential for biotechnological application, especially lignocellulose biomass hydrolysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Xylanase aus Trichoderma viride, lyophilized powder, ≥100 units/mg protein