Merck
The dietary acrylamide intake adversely affects the serum trace element status.
Biological trace element research (2013-01-12)
Fatma Hümeyra Yerlikaya, Yeşim Yener
ZUSAMMENFASSUNG
Acrylamide is an organic chemical which occurs in foods widespreadly consumed in diets worldwide. The purpose of this study was to evaluate the serum trace element levels (Fe, Cu, Zn, Mn, Cr, Se, Co, Ni, V, As, Mg, P, Li, K, Al) in Wistar rats exposed to acrylamide. Acrylamide was administered to the treatment groups at 2 and 5 mg/kg body weight (bw)/day via drinking water for 90 days. Inductively coupled plasma mass spectrometry was used for the determination of serum trace element concentrations. Serum Zn, Se, Co, V and Mg concentrations of 5 mg/kg bw/day acrylamide-treated male rats were lower, whereas serum As concentration was higher than the same parameters of the controls rats. Similarly, serum Zn, Se, Co, V and Mg concentrations were decreased in 5 mg/kg bw/day acrylamide-treated female rats compared with control rats. On the other hand, there were no significant differences between serum Fe, Cu, Mn, Cr, Ni, P, Li, K and Al concentrations of all groups. The results from this study provide evidence that dietary acrylamide intake adversely affects the serum trace elements status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamid, analytical standard
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Acrylamid, purum, ≥98.0% (GC)
Supelco
Acrylamid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acrylamid -Lösung, 40% in H2O
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Acrylamid, for Northern and Southern blotting, powder blend