Merck
  • Immobilized graphene-based composite from asphalt: facile synthesis and application in water purification.
Immobilized graphene-based composite from asphalt: facile synthesis and application in water purification.
Journal of hazardous materials (2013-01-15)
Theruvakkattil Sreenivasan Sreeprasad, Soujit Sen Gupta, Shihabudheen Mundampra Maliyekkal, Thalappil Pradeep
ZUSAMMENFASSUNG
An in situ strategy for the preparation of graphene immobilized on sand using asphalt, a cheap carbon precursor is presented. The as-synthesized material was characterized in detail using various spectroscopic and microscopic techniques. The presence of G and D bands at 1578 cm(-1) and 1345 cm(-1) in Raman spectroscopy and the 2D sheet-like structure with wrinkles in transmission electron microscopy confirmed the formation of graphenic materials. In view of the potential applicability of supported graphenic materials in environmental application, the as-synthesized material was tested for purifying water. Removal of a dye (rhodamine-6G) and a pesticide (chlorpyrifos), two of the important types of pollutants of concern in water, were investigated in this study. Adsorption studies were conducted in batch mode as a function of time, particle size, and adsorbent dose. The continuous mode experiments were conducted in multiple cycles and they confirmed that the material can be used for water purification applications. The adsorption efficacy of the present adsorbent system was compared to other reported similar adsorbent systems and the results illustrated that the present materials are superior. The adsorbents were analyzed for post treatment and their reusability was evaluated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rhodamin 6G, Dye content ~95 %
Supelco
Chlorpyriphos, PESTANAL®, analytical standard
Sigma-Aldrich
Rhodamin 6G, Dye content 99 %
Sigma-Aldrich
Rhodamin 6G, suitable for fluorescence, BioReagent
Supelco
Chlorpyriphos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Rhodamin 6G -perchlorat, Dye content 99 %
Supelco
Rhodamin 6G, analytical standard