Merck
Far-infrared optical conductivity of CeCu2Si2.
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-15)
J Sichelschmidt, A Herzog, H S Jeevan, C Geibel, F Steglich, T Iizuka, S Kimura
ZUSAMMENFASSUNG
We investigated the optical reflectivity of the heavy-fermion metal CeCu(2)Si(2) in the energy range 3 meV-30 eV for temperatures between 4 and 300 K. The results for the charge dynamics indicate a behavior that is expected for the formation of a coherent heavy quasiparticle state: upon cooling the spectra of the optical conductivity indicate a narrowing of the coherent response. Below temperatures of 30 K a considerable suppression of conductivity evolves below a peak structure at 13 meV. We assign this gap-like feature to strong electron correlations due to the 4f-conduction electron hybridization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cäsium, ingot, ≥99.95% trace metals basis