Merck
Crystallization and dissolution behavior of naproxen/polyethylene glycol solid dispersions.
The journal of physical chemistry. B (2013-01-17)
Qing Zhu, Scott J Toth, Garth J Simpson, Hsin-Yun Hsu, Lynne S Taylor, Michael T Harris
ZUSAMMENFASSUNG
The crystallization kinetics of naproxen (NAP) in NAP/polyethylene glycol (NAP/PEG) solid dispersions prepared at different crystallization temperatures was studied by in situ small-angle X-ray scattering/wide-angle X-ray scattering (SAXS/WAXS). It was found that the crystallization rate of NAP was faster at 25 °C in comparison to 40 °C. This resulted in different sizes of NAP domains, and consequently impacted the dissolution behavior. The sizes of NAP domains prepared at 40 °C were larger than those at 25 °C, as determined with surface area analysis, utilizing second-order nonlinear optical imaging of chiral crystals (SONICC). Consistent with this observation, the corresponding dissolution rate of the NAP/PEG dispersion prepared at 40 °C was indeed slower than that prepared at 25 °C. The microstructure of the NAP/PEG solid dispersions and the dissolution behavior also showed a dependence on the chemical composition of the solid dispersions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthalinessigsäure, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard