Merck
  • Acidic infusion in early reperfusion affects the activity of antioxidant enzymes in postischemic isolated rat heart.
Acidic infusion in early reperfusion affects the activity of antioxidant enzymes in postischemic isolated rat heart.
The Journal of surgical research (2013-01-22)
Claudia Penna, Maria-Giulia Perrelli, Francesca Tullio, Carmelina Angotti, Pasquale Pagliaro
ZUSAMMENFASSUNG
Acidic perfusion (AP) performed at the onset of reperfusion (i.e., acid postconditioning) is cardioprotective. We investigated the effect of AP on postischemic cardiac function and on the activity of endogenous superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase. The role of exogenous CAT or SOD on AP cardioprotection was also investigated. Phosphorylation of redox-sensitive survival kinases (protein kinase C [PKC] ε and extracellular signal-regulated kinase [ERK] 1/2) was also checked. Isolated rat hearts underwent ischemia and reperfusion (I/R) for 30 and 120 min, respectively. AP was obtained by lowering [HCO3(-)] in the perfusion buffer. Infarct size and left ventricular pressure were measured. Protocols include I/R only, I/R plus acidic perfusion in early reperfusion (I/R + AP), and I/R plus AP and CAT (I/R + AP + CAT) or SOD (I/R + AP + SOD). I/R + SOD and I/R + CAT additional hearts served as controls. AP and/or antioxidants were given in the initial 3 min of reperfusion. Enzyme activities were studied in postischemic phase (seventh minute of reperfusion) in I/R or I/R + AP and Sham (buffer-perfused) hearts. AP with (I/R + AP + CAT or I/R + AP + SOD) or without (I/R + AP) antioxidant enzymes resulted in a larger reduction of infarct size compared with I/R, I/R + SOD, or I/R + CAT. Compared with I/R, the postischemic systolic and diastolic recoveries of the cardiac function were markedly improved by the addition of AP and a lesser extent by AP + SOD or AP + CAT. AP increased the postischemic activity of CAT and lowered that of SOD and glutathione peroxidase compared with I/R only. Also, the phosphorylation and activity of ERK1/2 and PKCε were increased by AP. Acid postconditioning affects the activity of endogenous antioxidant enzymes, activates ERK1/2-PKCε pathways, and protects against myocardial I/R injury. The combination of AP and exogenous SOD or CAT still provides cardioprotection. It is likely that intracellular (not extracellular) redox condition plays a pivotal role in acidic protection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Supelco
Trizma® Base, reference material for titrimetry, certified by BAM, >99.5%
SAFC
Tromethamin