Merck
  • Occurrence of alternariol and alternariol monomethyl ether in beverages from the Entre Rios Province market, Argentina.
Occurrence of alternariol and alternariol monomethyl ether in beverages from the Entre Rios Province market, Argentina.
Mycotoxin research (2013-01-22)
Leticia Broggi, Cora Reynoso, Silvia Resnik, Fernanda Martinez, Vanesa Drunday, Angela Romero Bernal
ZUSAMMENFASSUNG
One hundred and eighty five samples of red, white and rosé wines and different juices purchased in Entre Rios, Argentina, were analyzed for the Alternaria mycotoxins alternariol (AOH) and alternariol methyl ether (AME). White wines were analyzed after removal of alcohol by a nitrogen stream and concentrated. AOH in red wines was cleaned up by solid-phase extraction columns in series (octadecyl and amino propyl modified silica) and AME quantified directly on the sample. The juices were filtered and concentrated, and then all sample extracts were quantified by high performance liquid chromatography with photodiode array detector that allows confirmation through UV spectra. Method validation revealed a good sensitivity with adequate LOD and LOQ for AME and less sensitivity for AOH (i.e. white wine: AME 0.8 and 1.4 ng/mL, AOH 2 and 3.3 ng/mL; red wine: AME 0.1 and 0.2 ng/mL, AOH 4.5 and 7.5 ng/mL; apple juice: AME 1.7 and 2.8 ng/mL, AOH 5 and 9 ng/mL; other juices: AME 2.0 and 3.1 ng/mL, AOH 6 and 10 ng/mL). Recoveries in all cases were greater than 80 %. Four of 53 white wine samples were contaminated with AOH with a maximum level of 18 ng/mL, 6 of 56 samples of red wine had a maximum of 13 ng/mL, and 3 of 68 samples of juices had traces of AOH. AME was less frequently detected than AOH, and the LOD and LOQ for AME are smaller than for AOH. Only three samples of white wine and one of red wine were contaminated, but in only one white wine sample (AME 225 ng/mL) did the toxin level exceed the LOQ.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Alternariol from Alternaria sp., ~96%
Supelco
Alternariol, analytical standard