Merck
  • Engineering a cyanobacterium as the catalyst for the photosynthetic conversion of CO2 to 1,2-propanediol.
Engineering a cyanobacterium as the catalyst for the photosynthetic conversion of CO2 to 1,2-propanediol.
Microbial cell factories (2013-01-24)
Han Li, James C Liao
ZUSAMMENFASSUNG
The modern society primarily relies on petroleum and natural gas for the production of fuels and chemicals. One of the major commodity chemicals 1,2-propanediol (1,2-PDO), which has an annual production of more than 0.5 million tons in the United States, is currently produced by chemical processes from petroleum derived propylene oxide, which is energy intensive and not sustainable. In this study, we sought to achieve photosynthetic production of 1,2-PDO from CO2 using a genetically engineered cyanobacterium Synechococcus elongatus PCC 7942. Compared to the previously reported biological 1,2-PDO production processes which used sugar or glycerol as the substrates, direct chemical production from CO2 in photosynthetic organisms recycles the atmospheric CO2 and will not compete with food crops for arable land. In this study, we reported photosynthetic production of 1,2-PDO from CO2 using a genetically engineered cyanobacterium Synechococcus elongatus PCC 7942. Introduction of the genes encoding methylglyoxal synthase (mgsA), glycerol dehydrogenase (gldA), and aldehyde reductase (yqhD) resulted in the production of ~22 mg/L 1,2-PDO from CO2. However, a comparable amount of the pathway intermediate acetol was also produced, especially during the stationary phase. The production of 1,2-PDO requires a robust input of reducing equivalents from cellular metabolism. To take advantage of cyanobacteria's NADPH pool, the synthetic pathway of 1,2-PDO was engineered to be NADPH-dependent by exploiting the NADPH-specific secondary alcohol dehydrogenases which have not been reported for 1,2-PDO production previously. This optimization strategy resulted in the production of ~150 mg/L 1,2-PDO and minimized the accumulation of the incomplete reduction product, acetol. This work demonstrated that cyanobacteria can be engineered as a catalyst for the photosynthetic conversion of CO2 to 1,2-PDO. This work also characterized two NADPH-dependent sADHs for their catalytic capacity in 1,2-PDO formation, and suggested that they may be useful tools for renewable production of reduced chemicals in photosynthetic organisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
(S)-(+)-1,2-Propandiol, 96%
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propandiol, analytical standard
Sigma-Aldrich
Glycerin-Dehydrogenase aus Cellulomonas sp., lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
(R)-(−)-1,2-Propandiol, 96%
Sigma-Aldrich
1,2-Propandiol, tested according to Ph. Eur.