Merck
  • Removal of Cr(VI) from aqueous solution by flocculant with the capacity of reduction and chelation.
Removal of Cr(VI) from aqueous solution by flocculant with the capacity of reduction and chelation.
Journal of hazardous materials (2013-01-24)
Gang Wang, Qing Chang, Xiaoting Han, Mingyue Zhang
ZUSAMMENFASSUNG
A novel agent polyethyleneimine-sodium xanthogenate (PEX) with the multifunction of reduction, chelation, flocculation and precipitation was synthesized by using polyethyleneimine (PEI), carbon disulfide (CS2), and sodium hydroxide (NaOH). The effects of different important parameters, such as pH value, initial Cr(VI) concentration, coexisting ions and turbidity etc., on the removal of chromium from aqueous solution by PEX were investigated in flocculation experiments. The experiments results demonstrated that PEX could efficiently remove Cr(VI) and total Cr (Cr(VI)+Cr(III)) in strongly acidic media. It was proved that the presence of coexisting ions (Na(+), Ca(2+), F(-), Cl(-), and SO4(2-)) in the solution had a little influence on the removal of chromium. Furthermore, it was conformed that Cr(VI) ions and turbidity could be simultaneously removed when water samples contained both Cr(VI) ions and turbidity. Finally, the mechanism of interaction between chromium and PEX was further confirmed by Fourier transform infrared spectroscopy (FTIR) and scanning electron microscopy (SEM). The results reveal that dithiocarboxylic acid groups on PEX macromolecule play a major role in Cr(VI) reduction and Cr(III) chelation, and the flocs formation is attributed to the interparticle bridging mechanism of PEX.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Kohlenstoffdisulfid, ACS reagent, ≥99.9%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Kohlenstoffdisulfid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Kohlenstoffdisulfid, ACS reagent, ≥99.9%
Supelco
Natronlauge, 49-51% in water, eluent for IC
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Kohlenstoffdisulfid, anhydrous, ≥99%
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Kohlenstoffdisulfid -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Natriumhydroxid, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets