- Primary jejunal adenocarcinoma incidentally diagnosed on positron emission tomography/computed tomography in a patient with metastatic colorectal cancer: suspicion of Lynch syndrome and effect on therapeutic management.
Primary jejunal adenocarcinoma incidentally diagnosed on positron emission tomography/computed tomography in a patient with metastatic colorectal cancer: suspicion of Lynch syndrome and effect on therapeutic management.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-01-24)
Wulfran Cacheux, Astrid Lièvre, Pascale Mariani, Vincent Servois, Fereshteh Farkhondeh, Etienne Rouleau, Bruno Buecher, Slavomir Petras
PMID23341528