Merck
Chemical constituents of three Allium species from Romania.
Molecules (Basel, Switzerland) (2013-01-25)
Laurian Vlase, Marcel Parvu, Elena Alina Parvu, Anca Toiu
ZUSAMMENFASSUNG
The aim of this work was to study the chemical composition of Allium obliquum L., A. senescens L. subsp. montanum (Fries) Holub, and A. schoenoprasum L. subsp. schoenoprasum. Sulphur-containing compounds analysis was performed by an LC-MS method, the identification and quantification of polyphenolic compounds through a HPLC-UV-MS method, and the presence of five sterols was simultaneously assessed by HPLC-MS-MS. Alliin was identified only in A. obliquum and A. senescens subsp. montanum extracts, whilst allicin was present in all extracts, with higher amounts in A. schoenoprasum and A. obliquum. The pattern of phenol carboxylic acids shows the presence of p-coumaric and ferulic acids in all species. Isoquercitrin was identified in A. obliquum and A. schoenoprasum, and rutin in A. senescens subsp. montanum and A. schoenoprasum. Luteolin and apigenin were identified only in A. obliquum. All three species contain glycosides of kaempferol and quercetol. β-Sitosterol and campesterol were identified in all species. The results obtained showed significant differences in the composition of the three Allium species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Supelco
Kaempferol, analytical standard