Merck
  • Human Vγ9Vδ2-T cells efficiently kill influenza virus-infected lung alveolar epithelial cells.
Human Vγ9Vδ2-T cells efficiently kill influenza virus-infected lung alveolar epithelial cells.
Cellular & molecular immunology (2013-01-29)
Hong Li, Zheng Xiang, Ting Feng, Jinrong Li, Yinping Liu, Yingying Fan, Qiao Lu, Zhongwei Yin, Meixing Yu, Chongyang Shen, Wenwei Tu
ZUSAMMENFASSUNG
γδ-T cells play an indispensable role in host defense against different viruses, including influenza A virus. However, whether these cells have cytotoxic activity against influenza virus-infected lung alveolar epithelial cells and subsequently contribute to virus clearance remains unknown. Using influenza virus-infected A549 cells, human lung alveolar epithelial cells, we investigated the cytotoxic activity of aminobisphosphonate pamidronate (PAM)-expanded human Vγ9Vδ2-T cells and their underlying mechanisms. We found that PAM could selectively activate and expand human Vγ9Vδ2-T cells. PAM-expanded human Vγ9Vδ2-T cells efficiently killed influenza virus-infected lung alveolar epithelial cells and inhibited virus replication. The cytotoxic activity of PAM-expanded Vγ9Vδ2-T cells was dependent on cell-to-cell contact and required NKG2D activation. Perforin-granzyme B, tumor-necrosis factor-related apoptosis-inducing ligand (TRAIL) and Fas-Fas ligand (FasL) pathways were involved in their cytotoxicity. Our study suggests that targeting γδ-T cells by PAM can potentially offer an alternative option for the treatment of influenza virus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pamidronate disodium salt hydrate, ≥95% (NMR), solid