Merck
Analysis of transplanted bone marrow-derived cells in chronic pancreatitis.
Methods in molecular biology (Clifton, N.J.) (2013-01-30)
Christoph B Westphalen, Frederic Marrache, Timothy C Wang
ZUSAMMENFASSUNG
Pancreatic cancer is a uniformly lethal disease characterized by a strong stromal reaction called desmoplasia. Organ fibrosis is also a feature of chronic pancreatitis a known risk factor for pancreatic cancer. Here we describe a transplantation approach to investigate bone marrow-derived cells in murine models of chronic pancreatitis and pancreatic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caerulein, ≥95% (HPLC)