- [Research progress studies on pharmacology and pharmacokinetics of ligustilide].
[Research progress studies on pharmacology and pharmacokinetics of ligustilide].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-02-05)
Ai-Hua Zuo, Li Wang, Hong-Bin Xiao
PMID23373200
ZUSAMMENFASSUNG
Ligustilide is contained highly or around 1% in such umbelliferous plants as Angelica sinensis and Ligusticum chuanxiong, is one of main bioactive constituents. It shows many pharmacological activities related to their efficacy. At present, ligustilide has attracted extensive attention and more and more studies have been reported, indicating that it is a promising compound. This essay summarizes the progress of pharmacological effects of ligustilide on neuroprotection, vasodilatation, anti-caner and anti-tumor, analgesia and anti-inflammation, and pharmacokinetics including absorption, distribution, metabolism and excretion, providing basis for further studies and development of ligustilide.