- Tetrathiafulvalene-capped hybrid materials for the optical detection of explosives.
Tetrathiafulvalene-capped hybrid materials for the optical detection of explosives.
ACS applied materials & interfaces (2013-02-05)
Yolanda Salinas, Ramón Martínez-Máñez, Jan O Jeppesen, Lars H Petersen, Félix Sancenón, María Dolores Marcos, Juan Soto, Carmen Guillem, Pedro Amorós
PMID23373746
ZUSAMMENFASSUNG
Mesoporous silica microparticles capped with TTF moieties and containing a ruthenium dye in the pores were used for the turn-on optical detection of the nitroaromatic explosives Tetryl and TNT via a selective pore uncapping and release of the entrapped dye.