Merck
Molecular dynamics of the interaction of anionic surfactants with liposomes.
Colloids and surfaces. B, Biointerfaces (2013-02-05)
Changfeng Chen, Cuihong Jiang, Carl P Tripp
ZUSAMMENFASSUNG
The time dependent molecular interactions of sodium dodecyl sulfate (SDS) with dipalmitoylphosphatidylcholine (DPPC) liposomes were studied using attenuated total reflection-Fourier transform infrared (ATR-FTIR) spectroscopy. In particular, the ATR spectra provided information on the dynamic amount of SDS incorporated and DPPC expelled in the DPPC structure along with structural information on the molecular nature of the SDS/DPPC assemblies. It was found that the uptake of SDS was always mirrored by the ejection of DPPC molecules from the liposomes and that the incorporation of SDS followed two routes; SDS intercalated into the DPPC liposomes and SDS was bound electrostatically to the charged surface of the DPPC liposome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphocholine, ~99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri