Merck
  • Combined treatment of earlobe keloids with shaving, cryosurgery, and intralesional steroid injection: a 1-year follow-up.
Combined treatment of earlobe keloids with shaving, cryosurgery, and intralesional steroid injection: a 1-year follow-up.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2013-02-06)
Mariana Figueiroa Careta, Ana C Fortes, Maria Cristina Messina, Celina W Maruta
ZUSAMMENFASSUNG
Earlobe keloids are benign, fibrous proliferations that occur in predisposed persons at sites of cutaneous injury. No single best therapeutic modality is indicated. To describe a 1-year follow-up of 12 patients with earlobe keloids treated by shaving followed by cryosurgery and intralesional injection of triamcinolone. Twelve patients were treated with combined surgery and cryosurgery. After 1 year, major response was observed in nine cases (75%) and moderate response in two cases (16%); one case had a relapse 5 months after the surgery. These results are highly encouraging because all patients showed improvement. Shaving associated with cryosurgery seems to be a useful treatment for large keloids scars.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triamcinolon