Merck
Chemical enhancement of footwear impressions in blood on fabric - part 3: amino acid staining.
Science & justice : journal of the Forensic Science Society (2013-02-06)
Kevin J Farrugia, Helen Bandey, Kathleen Savage, Niamh NicDaéid
ZUSAMMENFASSUNG
Enhancement of footwear impressions, using ninhydrin or ninhydrin analogues is not considered common practice and such techniques are generally used to target amino acids present in fingermarks where the reaction gives rise to colour and possibly fluorescence. Ninhydrin and two of its analogues were used for the enhancement of footwear impressions in blood on various types, colours and porosities of fabric. Test footwear impressions on fabric were prepared using a specifically built rig to minimise the variability between each impression. Ninhydrin enhancement of footwear impressions in blood on light coloured fabric yielded good enhancement results, however the contrast was weak or non-existent on dark coloured fabrics. Other ninhydrin analogues which have the advantage of fluorescence failed to enhance the impressions in blood on all fabrics. The sequential treatment of impressions in blood on fabric with other blood enhancing reagents (e.g. protein stains and heme reagents) was also investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection