Merck
Analysis of light scattering by red blood cells in ektacytometry using global pattern fitting.
Biorheology (2013-02-06)
Miklos Rabai, Herbert J Meiselman, Rosalinda B Wenby, Jon A Detterich, Jack Feinberg
ZUSAMMENFASSUNG
Ektacytometry measures the shape of red blood cells under shear stress by analyzing the diffraction pattern of laser light passing through a thin layer of suspended cells. Here we model the diffraction pattern using a combination of Bessel and anomalous scattering functions, and employ a global pattern-fitting technique over nine different shear stresses to determine the separate mechanical properties of normal and non-deformable cells. This technique is capable of yielding the correct elongation index of the normal cells over a range of shear stresses even when they are mixed with as much as 50% non-deformable cells. Additionally, the relative concentrations of normal and non-deformable cells can be determined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications