Merck
  • Structure of ALD1, a plant-specific homologue of the universal diaminopimelate aminotransferase enzyme of lysine biosynthesis.
Structure of ALD1, a plant-specific homologue of the universal diaminopimelate aminotransferase enzyme of lysine biosynthesis.
Acta crystallographica. Section F, Structural biology and crystallization communications (2013-02-07)
Vladimir Sobolev, Marvin Edelman, Orly Dym, Tamar Unger, Shira Albeck, Menny Kirma, Gad Galili
ZUSAMMENFASSUNG
Diaminopimelate aminotransferase (DAP-AT) is an enzyme in the lysine-biosynthesis pathway. Conversely, ALD1, a close homologue of DAP-AT in plants, uses lysine as a substrate in vitro. Both proteins require pyridoxal-5'-phosphate (PLP) for their activity. The structure of ALD1 from the flowering plant Arabidopsis thaliana (AtALD1) was solved at a resolution of 2.3 Å. Comparison of AtALD1 with the previously solved structure of A. thaliana DAP-AT (AtDAP-AT) revealed similar interactions with PLP despite sequence differences within the PLP-binding site. However, sequence differences between the binding site of AtDAP-AT for malate, a purported mimic of substrate binding, and the corresponding site in AtALD1 led to different interactions. This suggests that either the substrate itself, or the substrate-binding mode, differs in the two proteins, supporting the known in vitro findings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Diamino-pimelinsäure, ≥98% (TLC)
Sigma-Aldrich
2,6-Diamino-pimelinsäure, ≥95.0% (NT)