Merck
  • An aide memoire for model-based drug development--a new key for the interpretation of the erythromycin breath test--therapeutic index as a biomarker for smart drug development.
An aide memoire for model-based drug development--a new key for the interpretation of the erythromycin breath test--therapeutic index as a biomarker for smart drug development.
Biomarkers in medicine (2013-02-08)
Bruno Boutouyrie-Dumont
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin-Standard -Lösung, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard