Merck
  • Distal pup cues evoke dopamine responses in hormonally primed rats in the absence of pup experience or ongoing maternal behavior.
Distal pup cues evoke dopamine responses in hormonally primed rats in the absence of pup experience or ongoing maternal behavior.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-02-09)
Veronica M Afonso, Waqqas M Shams, Daniel Jin, Alison S Fleming
ZUSAMMENFASSUNG
During the early postpartum period or following estrogen/progesterone administration, pups elicit maternal behavior accompanied by a robust dopamine (DA) response in the nucleus accumbens (NAC) of female rats (Afonso et al., 2009). To determine whether DA responds to ostensibly "salient" stimuli in the absence of consummatory behaviors, we examined NAC shell DA responses during restricted (stimuli placed in a perforated box), and unrestricted access to pup and food stimuli. Microdialysis samples were collected from female rats that were either cycling and postpartum (Experiment 1), or after ovariectomy and treated with empty and hormone-filled capsules (Experiment 2). Relative to nonprimed controls, hormonally primed females had suppressed basal DA concentrations and facilitated pup-evoked DA responses, regardless of stimulus access condition. In contrast, food-evoked DA responses were unchanged by hormonal priming and were greater when females consumed food compared with distal (restricted) exposure to food. During pup and food restriction conditions, the lack of any "appetitive" behavioral differences, even in pup experienced postpartum females, was surprising. In Experiment 3, we confirmed that postpartum dams allocated time equivalently to restricted pup and food stimuli, even after pup deprivation. This was in sharp contrast to the effects of deprivation during the unrestricted access phase. Together, our data demonstrated that, in hormonally primed females, distal pup cues could evoke DA responses without prior stimulus experience, ongoing maternal (behavioral) responses, or clear evidence of robust pup saliency. The results suggest that NAC DA response reflects a state of responsiveness related to basal DA suppression in the hormonally primed female rat.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Oestradiol-3-benzoat, ≥97%