Merck
Liver kinase b1 is required for white adipose tissue growth and differentiation.
Diabetes (2013-02-12)
Wencheng Zhang, Qilong Wang, Ping Song, Ming-Hui Zou
ZUSAMMENFASSUNG
White adipose tissue (WAT) is not only a lipogenic and fat storage tissue but also an important endocrine organ that regulates energy homeostasis, lipid metabolism, appetite, fertility, and immune and stress responses. Liver kinase B1 (LKB1), a tumor suppressor, is a key regulator in energy metabolism. However, the role of LKB1 in adipogenesis is unknown. The current study aimed to determine the contributions of LKB1 to adipogenesis in vivo. Using the Fabp4-Cre/loxP system, we generated adipose tissue-specific LKB1 knockout (LKB1(ad-/-)) mice. LKB1(ad-/-) mice exhibited a reduced amount of WAT, postnatal growth retardation, and early death before weaning. Further, LKB1 deletion markedly reduced the levels of insulin receptor substrate 1 (IRS1), peroxisome proliferator-activated receptor γ, CCAAT/enhancer-binding protein α, and phosphorylated AMP-activated protein kinase (AMPK). Consistent with these results, overexpression of constitutively active AMPK partially ablated IRS1 degradation in LKB1-deficient cells. LKB1 deletion increased the levels of F-box/WD repeat-containing protein (Fbw) 8, the IRS1 ubiquitination E3 ligase. Silencing of Fbw8 increased IRS1 levels. Finally, promoter analysis and DNA chromatin immunoprecipitation analysis identified three sterol regulatory element (SRE) sites in the Fbw8 promoter, where SRE-binding protein 1c binds and induces the expression of Fbw8. Taken together, these data indicate that LKB1 controls IRS1-dependent adipogenesis via AMPK in WAT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Myokinase aus Kaninchenmuskel, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)
Sigma-Aldrich
Chromatin-Immunpräzipitations-Kit Magna ChIP® A/G, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
Anti-IRS1-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-Uncoupling Protein 1 Antibody, Chemicon®, from rabbit