Merck
Targeted cancer therapy with a 2-deoxyglucose-based adriamycin complex.
Cancer research (2013-02-12)
Jie Cao, Sisi Cui, Siwen Li, Changli Du, Junmei Tian, Shunan Wan, Zhiyu Qian, Yueqing Gu, Wei R Chen, Guangji Wang
ZUSAMMENFASSUNG
Adriamycin (ADM) has been effective against many types of solid tumors in clinical applications. However, its use is limited because of systemic toxicities, primarily cardiotoxicity, and multidrug resistance. In this study, a new active receptor-mediated complex, ADM conjugated with 2-amino-2-deoxy-d-glucose and succinic acid (2DG-SUC-ADM), was designed to target tumor cells through glucose transporter 1 (GLUT1). MTT assay and confocal images showed that the complex had better inhibition rate to tumor cells and low toxicity to normal cells. Most importantly, the complex displayed a potential to reverse overcome multidrug resistance in cancer cells, with more complex transported into the nucleus of tumor cells. Our in vivo experiments also showed that this new complex could significantly decrease organ toxicity and enhance the antitumor efficacy compared with free ADM, indicating a promising drug of 2DG-SUC-ADM for targeted cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
Bernsteinsäure, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Supelco
Bernsteinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland