Merck
  • Molecular cloning, expression and regulation analysis of the interleukin-6 (IL-6) gene in goose adipocytes.
Molecular cloning, expression and regulation analysis of the interleukin-6 (IL-6) gene in goose adipocytes.
British poultry science (2013-02-13)
F Wang, Y Tian, G Li, X Chen, H Yuan, D Wang, J Li, J Shen, Z Tao, Y Fu, L Lu
ZUSAMMENFASSUNG
1. Interleukin-6 (IL-6) is a multifunctional cytokine involved in lipid metabolism in adipose tissue. The objective of the study was to identify and characterize the IL-6 gene in the goose. 2. A full-length coding sequence (CDS) of the goose (Anser anser) IL-6 gene was cloned that encoded a 234-amino acid peptide containing a 38-amino acid signal peptide, an IL-6/G-CSF/MGF family consensus pattern and four conserved α-helices. The mature goose IL-6 showed 74% and 39% identities to that of chicken and human, respectively. 3. Quantitative real-time PCR analysis showed that the goose IL-6 was predominantly expressed in liver and was up-regulated in adipocytes by lipopolysaccharide (LPS) and oleic acid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleinsäure, analytical standard
Supelco
Oleinsäure, Selectophore, ≥99%