Merck
  • Kinetics and mechanism of imidazole-catalyzed acylation of cellulose in LiCl/N,N-dimethylacetamide.
Kinetics and mechanism of imidazole-catalyzed acylation of cellulose in LiCl/N,N-dimethylacetamide.
Carbohydrate polymers (2013-02-13)
Haq Nawaz, Paulo Augusto R Pires, Omar A El Seoud
ZUSAMMENFASSUNG
Cellulose acylation by anhydrides (ethanoic to hexanoic) plus tosyl chloride, TsCl, or imidazole in LiCl/N,N-dimethylacetamide solution has been studied. Contrary to a previous claim, TsCl does not catalyze acylation. For the diazole-catalyzed reaction, N-acylimidazole is the acylating agent. Third order rate constants (k(3); 40-70 °C) have been calculated from conductivity data and split, by using information from model compounds, into contributions from the primary- (k(3;Prim(OH))) and secondary- (k(3;Sec(OH))) hydroxyl groups of cellulose. Values of k(3,Prim(OH))/k(3,Sec(OH)) are >1, and increase linearly as a function of increasing the number of carbon atoms of the acyl group. Rate constants and the degree of biopolymer substitution decrease on going from ethanoic- to butanoic-, then increase for pentanoic- and hexanoic anhydride, due to enthalpy/entropy compensation. Relative to the uncatalyzed reaction, the diazole-mediated one is associated with smaller enthalpy- and larger entropy of activation, due to difference of the acylating agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylacetamid, ReagentPlus®, 99%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
N,N-Dimethylacetamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamid, anhydrous, 99.8%
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Lithiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
p-Toluolsulfonylchlorid, ReagentPlus®, ≥99%
Supelco
Lithiumchlorid -Lösung, 1 M in ethanol
Sigma-Aldrich
N,N-Dimethylacetamid, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
p-Toluolsulfonylchlorid, reagent grade, ≥98%
Sigma-Aldrich
Imidazol -hydrochlorid
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Sigma-Aldrich
Lithiumchlorid, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Imidazol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Imidazol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylacetamid, spectrophotometric grade, ≥99%
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Supelco
N,N-Dimethylacetamid, analytical standard
Sigma-Aldrich
Imidazolylnatrium, technical grade
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis