Merck
  • Kinetics of dextran crosslinking by epichlorohydrin: a rheometry and equilibrium swelling study.
Kinetics of dextran crosslinking by epichlorohydrin: a rheometry and equilibrium swelling study.
Carbohydrate polymers (2013-02-13)
Zahraalsadat Emami Meybodi, Mohammad Imani, Mohammad Atai
ZUSAMMENFASSUNG
Curing behavior and gel properties of dextran are investigated during its crosslinking by epichlorohydrin (ECH) using rheometry and equilibrium swelling studies. Each ECH/anhydroglucose unit of different molar ratio (0.16-0.99) was added to a known volume of aqueous dextran solution (25%, w/w) in the presence of sodium hydroxide (1.2-3.6 M) as a catalyst. Gel point was investigated as a function of the reactants concentration. The rheological behavior of the curing gels was evaluated using a rheokinetic model. Swelling behavior of the cured, cylindrical specimens was also studied gravimetrically. Increasing NaOH and ECH concentrations significantly increased the crosslinking reaction rate leading to shorter gel points, which was attributed to more ionization of hydroxyl functional groups of dextran due to higher NaOH concentrations. Degree of swelling vs. the reaction time was decreased sharply as a direct function of increasing NaOH and ECH concentrations up to certain levels, which then leveled off. In contrast, higher NaOH concentrations increased the swelling ratio probably due to the alkaline induced degradation of dextran chains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
(±)-Epichlorhydrin, purum, ≥99% (GC)
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
(S)-(+)-Epichlorhydrin, 98%
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
(±)-Epichlorhydrin, analytical standard
Sigma-Aldrich
Natriumhydroxid, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets