Merck
  • Modeling studies: Adsorption of aniline blue by using Prosopis Juliflora carbon/Ca/alginate polymer composite beads.
Modeling studies: Adsorption of aniline blue by using Prosopis Juliflora carbon/Ca/alginate polymer composite beads.
Carbohydrate polymers (2013-02-13)
M Kumar, R Tamilarasan
ZUSAMMENFASSUNG
The research article describes the experimental and modeling study for the adsorptive removal of aniline blue dye (AB dye) from aqueous matrices using a Prosopis Juliflora modified carbon/Ca/alginate polymer bead as a low cost and eco-friendly adsorbent. The rate of adsorption was investigated under various experimental parameters such as contact time, adsorbent dose, dye concentration, pH and temperature. The kinetics, equilibrium and thermodynamic studies were assessed to find out the efficiency of the adsorption process. The equilibrium uptake capacity of the adsorption process was found with Freundlich and Langmuir adsorption isotherm equations and it was evaluated by dimensionless separation factor (R(L)). The dynamics of adsorption was predicted by pseudo-first order, pseudo-second order Lagergren's equation and intra particle diffusion model. Adsorption feasibility was assessed with thermodynamic parameters such as isosteric heat of adsorption (ΔH°), standard entropy (ΔS°) and Gibbs free energy (ΔG°) using VantHoff plot. The alginate bead was characterized with FTIR spectroscopy and Scanning Electron Microscopy (SEM).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumalginat, powder
Sigma-Aldrich
Natriumalginat
Sigma-Aldrich
Natriumalginat aus Braunalgen, Medium viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, low viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginsäure aus Braunalgen, powder
Sigma-Aldrich
Calciumalginat aus Braunalgen