Merck
  • Conductive magnetite nanoparticles accelerate the microbial reductive dechlorination of trichloroethene by promoting interspecies electron transfer processes.
Conductive magnetite nanoparticles accelerate the microbial reductive dechlorination of trichloroethene by promoting interspecies electron transfer processes.
ChemSusChem (2013-02-13)
Federico Aulenta, Simona Rossetti, Stefano Amalfitano, Mauro Majone, Valter Tandoi
ZUSAMMENFASSUNG
Playing your part: Conductive magnetite nanoparticles accelerate the microbial reductive dechlorination of trichloroethene (TCE), an ubiquitous and toxic subsurface contaminant. The stimulatory effect most likely results from the nanoparticles promoting the establishment of interspecies electron transfer (IET) processes between non-dechlorinating and dechlorinating microorganisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trichlorethylen, ACS reagent, ≥99.5%
Supelco
Trichlorethylen, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichlorethylen, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichlorethylen -Lösung, certified reference material, 5000 μg/mL in methanol