Merck
  • Ultrasound, pH, and magnetically responsive crown-ether-coated core/shell nanoparticles as drug encapsulation and release systems.
Ultrasound, pH, and magnetically responsive crown-ether-coated core/shell nanoparticles as drug encapsulation and release systems.
ACS applied materials & interfaces (2013-02-14)
Siu-Fung Lee, Xiao-Ming Zhu, Yi-Xiang J Wang, Shou-Hu Xuan, Qihua You, Wing-Hong Chan, Chi-Hin Wong, Feng Wang, Jimmy C Yu, Christopher H K Cheng, Ken Cham-Fai Leung
ZUSAMMENFASSUNG
Core@shell nanoparticles with superparamagnetic iron oxide core, mesoporous silica shell, and crown ether periphery were fabricated toward drug delivery and tumor cell imaging. By the concept of nanovalve based on supramolecular gatekeeper, stimuli-responsive drug delivery nanosystems Fe3O4@SiO2@meso-SiO2@crown ethers were synthesized by (i) modified solvothermal reaction; (ii) sol-gel reaction; and (iii) amide coupling reaction. The successful coupling of the dibenzo-crown ethers onto the mesoporous silica shell was confirmed by thermogravimetric analysis and Infrared spectroscopy. In this system, the "ON/OFF" switching of the gatekeeper supramolecules can be controlled by pH-sensitive intramolecular hydrogen bonding or electrostatic interaction (such as metal chelating). Biological evaluation of the nanoparticles renders them noncytotoxic and can be uptaken by L929 cells. In this work, the antitumor drug (doxorubicin) loading and release profiles which were studied by the UV/visible absorption spectroscopy. The mechanism involves the best-fit binding of crown ethers with cesium or sodium ions at different pH values with ultrasonic wave in phosphate buffered saline (PBS). Magnetic resonance imaging analysis of the particles reveals a high relaxivity, rendering them potentially useful theranostic agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Diethylether, analytical standard