Merck
CREBH determines the severity of sulpyrine-induced fatal shock.
PloS one (2013-02-15)
Naganori Kamiyama, Masahiro Yamamoto, Hiroyuki Saiga, Ji Su Ma, Jun Ohshima, Sakaaki Machimura, Miwa Sasai, Taishi Kimura, Yoshiyasu Ueda, Hisako Kayama, Kiyoshi Takeda
ZUSAMMENFASSUNG
Although the pyrazolone derivative sulpyrine is widely used as an antipyretic analgesic drug, side effects, including fatal shock, have been reported. However, the molecular mechanism underlying such a severe side effect is largely unclear. Here, we report that the transcription factor CREBH that is highly expressed in the liver plays an important role in fatal shock induced by sulpyrine in mice. CREBH-deficient mice were resistant to experimental fatal sulpyrine shock. We found that sulpyrine-induced expression of cytochrome P450 2B (CYP2B) family genes, which are involved in sulpyrine metabolism, in the liver was severely impaired in CREBH-deficient mice. Moreover, introduction of CYP2B in CREBH-deficient liver restored susceptibility to sulpyrine. Furthermore, ectopic expression of CREBH up-regulated CYP2B10 promoter activity, and in vivo knockdown of CREBH in wild-type mice conferred a significant resistance to fatal sulpyrine shock. These data demonstrate that CREBH is a positive regulator of CYP2B in response to sulpyrine administration, which possibly results in fatal shock.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Aminoantipyrin, reagent grade
Sigma-Aldrich
4-Aminoantipyrin, puriss. p.a., reag. Ph. Eur., ≥99%
Supelco
4-Aminoantipyrin, for spectrophotometric det. of H2O2 and phenols, ≥98.0%