Merck
  • Influence of membrane structure on the preparation of colloidal lipid dispersions by premix membrane emulsification.
Influence of membrane structure on the preparation of colloidal lipid dispersions by premix membrane emulsification.
International journal of pharmaceutics (2013-02-16)
Sonja Joseph, Heike Bunjes
ZUSAMMENFASSUNG
Since premix membrane emulsification was developed as alternative technique for the preparation of emulsions and solid lipid particles as carrier systems for lipophilic drugs, many types of membranes have been used in this preparation process. The purpose of this study was to evaluate the influence of different types of membranes on the processability of colloidal lipid dispersions using the premix membrane emulsification technique. Polycarbonate, polyester, nylon, polyvinylidene fluoride, polysulfone and polyethersulfone membranes with a pore size of 200 nm were used in a small- and a large-volume extrusion process to yield dispersions of highly concentrated (20%) nonpolar lipids (medium chain triglycerides, trimyristin) stabilized by sodium dodecyl sulfate (15%) in aqueous solution. The structure of the different membranes was characterized by scanning electron microscopy. Similar mean particle sizes and size distributions were observed for all solid lipid nanoparticles obtained from both extrusion processes after 11 or 21 extrusion cycles. Smaller mean particle sizes were observed for emulsions prepared with track-etched membranes (polycarbonate, polyester). The use of a stack of four polycarbonate membranes in the large-volume extrusion process led to small particle sizes with monomodal size distributions already after one membrane passage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri