Merck
  • Sodium citrate and potassium phosphate as alternative adsorption buffers in hydrophobic and aromatic thiophilic chromatographic purification of plasmid DNA from neutralized lysate.
Sodium citrate and potassium phosphate as alternative adsorption buffers in hydrophobic and aromatic thiophilic chromatographic purification of plasmid DNA from neutralized lysate.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-02-16)
Nemailla Bonturi, Vanessa Soraia Cortez Oliveira Radke, Sônia Maria Alves Bueno, Sindélia Freitas, Adriano Rodrigues Azzoni, Everson Alves Miranda
ZUSAMMENFASSUNG
The number of studies on gene therapy using plasmid vectors (pDNA) has increased in recent years. As a result, the demand for preparations of pDNA in compliance with recommendations of regulatory agencies (EMEA, FDA) has also increased. Plasmid DNA is often obtained through fermentation of transformed Escherichia coli and purification by a series of unit operations, including chromatography. Hydrophobic interaction chromatography (HIC) and thiophilic aromatic chromatography (TAC), both using ammonium sulfate buffers, are commonly employed with success. This work was aimed at studying the feasibility of utilizing alternative salts in the purification of pDNA from neutralized lysate with phenyl-agarose (HIC) and mercaptopyrimidine-agarose (TAC) adsorbents. Their selectivity toward sc pDNA was evaluated through adsorption studies using 1.5 mol/L sodium citrate and 2.0 mol/L potassium phosphate as adsorption buffers. Chromatography with mercaptopyrimidine-agarose adsorbent and 1.5 mol/L sodium citrate was able to recover 91.1% of the pDNA with over 99.0% removal of gDNA and endotoxin. This represents a potential alternative for the primary recovery of sc pDNA. However, the most promising result was obtained using 2.0 mol/L potassium phosphate buffer and a mercaptopyrimidine-agarose column. In a single chromatographic step, this latter buffer/adsorbent system recovered 68.5% of the pDNA with 98.8% purity in accordance with the recommendations of regulatory agencies with regard to RNA and endotoxin impurity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Ammoniumsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, tribasisch, reagent grade, ≥97%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Kaliumphosphat, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Kaliumphosphat, reagent grade, ≥98.0%
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Kaliumphosphat -Lösung, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Ammoniumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, BioUltra, ≥99.0% (T)
Supelco
Ammoniumsulfat, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Kaliumphosphat, monobasisch -Lösung, reagent grade, 1.0 M in solution
Sigma-Aldrich
Kaliumphosphat, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ammoniumsulfat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumphosphat, 99.95% trace metals basis
Sigma-Aldrich
Ammoniumsulfat, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 Sulfat -Lösung, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N-sulfat -Lösung, 40 wt. % in H2O, 99.99 atom % 14N