Merck
  • Protective effect of marine algae phlorotannins against AAPH-induced oxidative stress in zebrafish embryo.
Protective effect of marine algae phlorotannins against AAPH-induced oxidative stress in zebrafish embryo.
Food chemistry (2013-02-16)
Min-Cheol Kang, Seon Heui Cha, W A J P Wijesinghe, Sung-Myung Kang, Seung-Hong Lee, Eun-A Kim, Choon Bok Song, You-Jin Jeon
ZUSAMMENFASSUNG
In this study the protective effect of phlorotannins, including phloroglucinol, eckol, dieckol, eckstolonol and triphloroethol A, isolated from brown algae Ecklonia cava was investigated against AAPH-induced oxidative stress toxicity in zebrafish embryos. Zebrafish embryos were exposed to AAPH and compared with other groups that were co-exposed with phlorotannins until 2-days post-fertilisation. All phlorotannins scavenged intracellular ROS and prevented lipid peroxidation and reduced AAPH-induced cell death in zebrafish embryos. Negative changes in morphological phenomena, such as pericardial oedema, yolk sac oedema, and growth retardation in zebrafish embryos exposed to AAPH were not observed in groups exposed to phlorotannins. These results clearly indicate that phlorotannins possess prominent antioxidant activity against AAPH-mediated toxicity and might be potential therapeutic agents for treating or preventing several diseases implicated with oxidative stress. This study provides a useful tool for examining the protective effect of antioxidants against AAPH-induced oxidative stress in an alternative in vivo model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2′-Azo-bis(2-methylpropionamidin) -dihydrochlorid, powder or granules, 97%
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
Phloroglucinol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.