Merck
  • Synthesis and evaluation of coumarin derivatives as potential dual-action HIV-1 protease and reverse transcriptase inhibitors.
Synthesis and evaluation of coumarin derivatives as potential dual-action HIV-1 protease and reverse transcriptase inhibitors.
Bioorganic & medicinal chemistry (2013-02-19)
Temitope O Olomola, Rosalyn Klein, Nicodemus Mautsa, Yasien Sayed, Perry T Kaye
ZUSAMMENFASSUNG
Baylis-Hillman-derived 3-(benzylaminomethyl)coumarins have been treated, sequentially, with chloroacetyl chloride and propargylamine to afford alkynylated coumarins as substrates for Click Chemistry reactions with azidothymidine (AZT) in the presence of a Cu(I) catalyst. The dual-action HIV-1 protease (PR) and reverse transcriptase (RT) inhibition potential of the resulting N-benzylated cycloaddition products, and a series of non-benzylated analogues, has been explored using saturation transfer difference (STD) NMR, computer modelling and enzyme inhibition techniques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3′-Azido-3′-Desoxythymidin, ≥98% (HPLC)
Supelco
Zidovudin, Pharmaceutical Secondary Standard; Certified Reference Material