Merck
  • Substituted piperidinyl glycinyl 2-cyano-4,5-methano pyrrolidines as potent and stable dipeptidyl peptidase IV inhibitors.
Substituted piperidinyl glycinyl 2-cyano-4,5-methano pyrrolidines as potent and stable dipeptidyl peptidase IV inhibitors.
Bioorganic & medicinal chemistry letters (2013-02-19)
Guohua Zhao, Chet Kwon, Aiying Wang, James G Robertson, Jovita Marcinkeviciene, Rex A Parker, Mark S Kirby, Lawrence G Hamann
ZUSAMMENFASSUNG
Synthesis and structure-activity relationship of a series of substituted piperidinyl glycine 2-cyano-4,5-methano pyrroline DPP-IV inhibitors are described. Improvement of the inhibitory activity and chemical stability of this series of compounds was respectively achieved by the introduction of bulky groups at the 4-position and 1-position of the piperidinyl glycine, leading to a series of potent and stable DPP-IV inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Piperidin, ReagentPlus®, 99%
Sigma-Aldrich
Piperidin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperidin, biotech. grade, ≥99.5%
Sigma-Aldrich
Piperidin -Lösung, suitable for peptide synthesis, 20% in DMF