Merck
Effects of radiation and vitamin C treatment on metronidazole genotoxicity in mice.
Mutation research (2013-02-19)
Lopamudra Das Roy, Sarbani Giri, Supriya Singh, Anirudha Giri
ZUSAMMENFASSUNG
The impact of exposure to low dose radiation (LDR) on human health is not clear. Besides, cross adaptation or sensitization with pharmaceutical agents may modify the risk of LDR. In the present study, we analyzed the interaction of radiation and metronidazole (MTZ) in inducing chromosome aberration (CA) and micronucleus (MN) in the bone marrow cells of Balb/C mice in vivo. Further, we evaluated the efficacy of vitamin C to reduce MTZ induced genotoxicity. We found that 10, 20 and 40mg/kg of MTZ induced dose dependent increase in the frequency of CA (r=0.9923, P<0.01) as well as MN (r=0.9823, P<0.05) in polychromatic erythrocytes. However, MTZ did not affect the ratio of polychromatic erythrocytes to normochromatic erythrocytes indicating lack of cytotoxicity. Supplementation with vitamin C prior to MTZ treatment significantly reduced the frequency of CA (P<0.001) as well as MN (P<0.001). Radiation (0.5Gy) exposure prior to MTZ treatment produced a less than additive (for CA) to additive (for MN) effects. However, radiation exposure following MTZ treatment produced additive (for CA) and synergistic (for MN) effects. Further, vitamin C pre-treatment also reduced the genotoxicity indices following the combined treatment of MTZ and radiation. Our findings suggest that MTZ may sensitize bone marrow cells to radiation exposure and enhances genotoxicity. We recommend more studies on the interaction of LDR and marketed pharmaceuticals to minimize possible harmful outcomes through appropriate precautionary measures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Metronidazol, BioXtra
Supelco
Metronidazol, analytical standard
Supelco
Metronidazol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metronidazol, VETRANAL®, analytical standard