Merck
  • Palladium(II) complexes as biologically potent metallo-drugs: synthesis, spectral characterization, DNA interaction studies and antibacterial activity.
Palladium(II) complexes as biologically potent metallo-drugs: synthesis, spectral characterization, DNA interaction studies and antibacterial activity.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-19)
Kollur Shiva Prasad, Linganna Shiva Kumar, Shivamallu Chandan, R M Naveen Kumar, Hosakere D Revanasiddappa
ZUSAMMENFASSUNG
Four novel mononuclear Pd(II) complexes have been synthesized with the biologically active Schiff base ligands (L1-L4) derived from 3-amino-2-methyl-4(3H)-quinazolinone. The structure of the complexes has been proposed by elemental analysis, molar conductance, IR, (1)H NMR, mass, UV-Vis spectrometric and thermal studies. The investigation of interaction of the complexes with calf thymus DNA (CT-DNA) has been performed with absorption and fluorescence spectroscopic studies. The nuclease activity was done using pUC19 supercoiled DNA by gel-electrophoresis. All the ligands and their Pd(II) complexes have also been screened for their antibacterial activity by discolor diffusion technique.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Desoxyribonukleinsäure aus Kalbsthymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Kalbsthymus, lyophilized powder
Sigma-Aldrich
Desoxyribonukleinsäure aus Kalbsthymus, Genomic, unsheared