Merck
Bacillus cereus fasciitis: a unique pathogen and clinically challenging sequela of inoculation.
American journal of orthopedics (Belle Mead, N.J.) (2013-02-23)
Andrew Rosenbaum, Dean Papaliodis, Maxwell Alley, Jordan Lisella, Michael Flaherty
ZUSAMMENFASSUNG
Bacillus cereus is an aerobic, spore-forming, gram-positive rod. It has historically been associated with "fried rice syndrome," a foodborne diarrheal and emetic illness resulting from eating fried rice dishes that have been sitting at room temperature for hours. We report the case of a 9-year-old boy who developed culture-positive B cereus fasciitis of the right lower extremity after being impaled on a tree branch. This case report further elucidates and emphasizes the importance of recognizing B cereus as a possible cause of severe soft-tissue infection. It must be included in the differential diagnosis of gas gangrene and necrotizing fasciitis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Clindamycin -hydrochlorid, lincosamide antibiotic
Supelco
Clindamycin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Clindamycin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard