- Acute oxalate nephropathy secondary to orlistat-induced enteric hyperoxaluria.
Acute oxalate nephropathy secondary to orlistat-induced enteric hyperoxaluria.
Nephrology (Carlton, Vic.) (2013-02-26)
Tony K Kwan, Steve J Chadban, Paul R McKenzie, John R Saunders
PMID23432752