Merck
  • A phase I/II pharmacokinetic and pharmacogenomic study of calcitriol in combination with cisplatin and docetaxel in advanced non-small-cell lung cancer.
A phase I/II pharmacokinetic and pharmacogenomic study of calcitriol in combination with cisplatin and docetaxel in advanced non-small-cell lung cancer.
Cancer chemotherapy and pharmacology (2013-02-26)
N Ramnath, S Daignault-Newton, G K Dy, J R Muindi, A Adjei, V L Elingrod, G P Kalemkerian, K B Cease, P J Stella, D E Brenner, S Troeschel, C S Johnson, D L Trump
ZUSAMMENFASSUNG
Preclinical studies demonstrated antiproliferative synergy of 1,25-D3 (calcitriol) with cisplatin. The goals of this phase I/II study were to determine the recommended phase II dose (RP2D) of 1,25-D3 with cisplatin and docetaxel and its efficacy in metastatic non-small-cell lung cancer. Patients were ≥18 years, PS 0-1 with normal organ function. In the phase I portion, patients received escalating doses of 1,25-D3 intravenously every 21 days prior to docetaxel 75 mg/m(2) and cisplatin 75 mg/m(2) using standard 3 + 3 design, targeting dose-limiting toxicity (DLT) rate <33 %. Dose levels of 1,25-D3 were 30, 45, 60, and 80 mcg/m(2). A two-stage design was employed for phase II portion. We correlated CYP24A1 tagSNPs with clinical outcome and 1,25-D3 pharmacokinetics (PK). 34 patients were enrolled. At 80 mcg/m(2), 2/4 patients had DLTs of grade 4 neutropenia. Hypercalcemia was not observed. The RP2D of 1,25-D3 was 60 mcg/m(2). Among 20 evaluable phase II patients, there were 2 confirmed, 4 unconfirmed partial responses (PR), and 9 stable disease (SD). Median time to progression was 5.8 months (95 % CI 3.4, 6.5), and median overall survival 8.7 months (95 % CI 7.6, 39.4). CYP24A1 SNP rs3787554 (C > T) correlated with disease progression (P = 0.03) and CYP24A1 SNP rs2762939 (C > G) trended toward PR/SD (P = 0.08). There was no association between 1,25-D3 PK and CYP24A1 SNPs. The RP2D of 1,25-D3 with docetaxel and cisplatin was 60 mcg/m(2) every 21 days. Pre-specified endpoint of 50 % confirmed RR was not met in the phase II study. Functional SNPs in CYP24A1 may inform future studies individualizing 1,25-D3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
1α,25-Dihydroxyvitamin D3 -Lösung, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Calcitriol, European Pharmacopoeia (EP) Reference Standard