Merck
  • Post-synthesis, characterization and catalytic properties of fluorine-planted MWW-type titanosilicate.
Post-synthesis, characterization and catalytic properties of fluorine-planted MWW-type titanosilicate.
Physical chemistry chemical physics : PCCP (2013-02-27)
Xiangqing Fang, Qiang Wang, Anmin Zheng, Yueming Liu, Longfei Lin, Haihong Wu, Feng Deng, Mingyuan He, Peng Wu
ZUSAMMENFASSUNG
F-Ti-MWW was post-synthesized by implanting fluorine species into a Ti-MWW framework through an acid treatment process in the presence of ammonium fluoride. The effects of NH4F addition amount, acid treatment temperature and precursor Ti content were investigated on the incorporation of F species, the zeolite structure and the coordination sites of Ti. Fluorine-implanting improved the surface hydrophobicity of the zeolite and altered the electropositivity nearby the tetrahedral Ti sites through forming the SiO3/2F and SiO4/2F(-) units. The negative effect of SiO4/2F(-) units in F-Ti-MWW was eliminated selectively by convenient anion-exchange with various alkali chlorides. F-Ti-MWW containing the SiO3/2F units possessed better catalytic activity and reusability, and a longer catalyst lifetime than conventional Ti-MWW.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumfluorid, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammoniumfluorid, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumhydrogendifluorid, reagent grade, 95%
Sigma-Aldrich
Ammoniumhydrogendifluorid, 99.999% trace metals basis