Merck
Polysaccharides from peach pulp: structure and effects on mouse peritoneal macrophages.
Food chemistry (2013-02-28)
F F Simas-Tosin, A P R Abud, C C De Oliveira, P A J Gorin, G L Sassaki, D F Bucchi, M Iacomini
ZUSAMMENFASSUNG
Pulp from peaches contained polygalacturonic acid and arabinogalactan as main polysaccharides, which were isolated and characterised. The polygalacturonic acid (AE-CWI) contained 95% GalA and its (13)C NMR spectrum showed signals at δ 98.9, 78.0, 71.4, 69.1, 68.4, and 175.1 from C-1, C-4, C-5, C-3, C-2, and C-6 respectively, from (1→4)-linked α-GalpA units. Methylation-MS analysis of carboxy-reduced material (AE-CWI-CR) gave 90% of 2,3,6-Me(3)-galactitol acetate. The arabinogalactan (AE-AG) was composed mainly of Ara (41%) and Gal (50%) and was characterised (methylation analysis and (13)C NMR) as a type II-arabinogalactan. It induced peritoneal macrophage activation in mice, ~70% of cells treated with this fraction (1-50 μg/mL) having morphology of activated cells. However, NO production in macrophages treated with AE-AG was not affected. This suggests a new biological activity for peach polysaccharides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-Arabinogalactan aus Lärchenholz, ≥80% (HPLC), from larch wood