Merck
Methylene blue for the treatment of refractory anaphylaxis without hypotension.
The American journal of emergency medicine (2013-03-05)
Paulo Roberto B Evora
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylenblau -Lösung, suitable for microscopy
Sigma-Aldrich
Methylenblau -Lösung, suitable for microbiology